Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC NH2-UAMC1110 TFA | 2990021-73-1 | 99.9% | 545.46 g·mol⁻¹ | C23H24F5N5O5 | 10 MG
SDP

Catalog No. 5000286102
Encompass_Preferred

NH2-UAMC1110 TFA is the trifluoroacetic acid salt of an amino-functionalized derivative of UAMC1110. It is used as a chemical intermediate for the synthesis of FAPI-QS and other FAP-targeting imaging agents and radiotracers. The compound is supplied with high purity and is intended for research and radiochemistry applications where an amino handle is required for conjugation and labeling.

  • High purity (99.89%) suitable for synthetic and radiochemistry use.
  • Amino-functionalized derivative for conjugation and labeling.
  • Trifluoroacetic acid salt form for improved handling and solubility.
  • Intended for use as an intermediate in FAP-targeting radiotracer synthesis.
  • Characterized by CAS number 2990021-73-1 and formula C23H24F5N5O5.

Catalog No. 50-002-86102 Supplier Medchemexpress LLC Supplier No. HY153552A10MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

NH2-UAMC1110 TFA is the trifluoroacetic acid salt of an amino-functionalized derivative of UAMC1110. It is used as a chemical intermediate for the synthesis of FAPI-QS and other FAP-targeting imaging agents and radiotracers. The compound is supplied with high purity and is intended for research and radiochemistry applications where an amino handle is required for conjugation and labeling.

  • High purity (99.89%) suitable for synthetic and radiochemistry use.
  • Amino-functionalized derivative for conjugation and labeling.
  • Trifluoroacetic acid salt form for improved handling and solubility.
  • Intended for use as an intermediate in FAP-targeting radiotracer synthesis.
  • Characterized by CAS number 2990021-73-1 and formula C23H24F5N5O5.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.