Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

[NH2Me2][(RuCl((R)-segphos(regR)))2(mu-Cl)3], TCI America™
SDP

Catalog No. N1021200MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
200 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N1021200MG 200 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N1021200MG Supplier TCI America Supplier No. N1021200MG
Add to Cart
Add to Cart

Chemical Identifiers

CAS 346457-41-8
Molecular Formula C78H67Cl5NO8P4Ru2
Molecular Weight (g/mol) 1649.68
MDL Number MFCD09753037
InChI Key GTKMQYLNAZIAEF-UHFFFAOYSA-M
Synonym Dimethylammonium Dichlorotri(mu-chloro)bis[(R)-(+)-5,5′C-bis(diphenylphosphino)-4,4′C-bi-1,3-benzodioxole]diruthenate(II)
PubChem CID 131674758
IUPAC Name [chloraniumyl(chloro)ruthenio]chloranium dimethylamine bis({4-[5-(diphenylphosphanyl)-2H-1,3-benzodioxol-4-yl]-2H-1,3-benzodioxol-5-yl}diphenylphosphane) hydrochloride ruthenium chloride
SMILES Cl.[Cl-].[Ru].CNC.[ClH+][Ru-]([ClH+])[ClH+].C1OC2=C(O1)C(=C(C=C2)P(C1=CC=CC=C1)C1=CC=CC=C1)C1=C(C=CC2=C1OCO2)P(C1=CC=CC=C1)C1=CC=CC=C1.C1OC2=C(O1)C(=C(C=C2)P(C1=CC=CC=C1)C1=CC=CC=C1)C1=C(C=CC2=C1OCO2)P(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Quantity 200 mg
Formula Weight 1646.65
Physical Form Crystalline Powder
Chemical Name or Material [NH2Me2][(RuCl((R)-segphos(regR)))2(mu-Cl)3]
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.