Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

[NH2Me2][(RuCl((S)-tolbinap))2(mu-Cl)3], TCI America™
SDP

Catalog No. N1018200MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N1018200MG 200 mg
N10181G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. N1018200MG Supplier TCI America Supplier No. N1018200MG
Add to Cart
Add to Cart

Chemical Identifiers

CAS 309735-86-2
Molecular Formula C98H88Cl5NP4Ru2
Molecular Weight (g/mol) 1783.07
MDL Number MFCD09753034
InChI Key FAAFCSOHMVBJIS-UHFFFAOYSA-J
Synonym Dimethylammonium Dichlorotri(mu-chloro)bis[(S)-(-)-2,2′C-bis(di-p-tolylphosphino)-1,1′C-binaphthyl]diruthenate(II)
PubChem CID 131675153
IUPAC Name bis(λ²-ruthenium(2+)) dimethylazanium bis({2'-[bis(4-methylphenyl)phosphanyl]-[1,1'-binaphthalen]-2-yl}bis(4-methylphenyl)phosphane) pentachloride
SMILES [Cl-].[Cl-].[Cl-].[Cl-].[Cl-].[Ru++].[Ru++].C[NH2+]C.CC1=CC=C(C=C1)P(C1=CC=C(C)C=C1)C1=CC=C2C=CC=CC2=C1C1=C2C=CC=CC2=CC=C1P(C1=CC=C(C)C=C1)C1=CC=C(C)C=C1.CC1=CC=C(C=C1)P(C1=CC=C(C)C=C1)C1=CC=C2C=CC=CC2=C1C1=C2C=CC=CC2=CC=C1P(C1=CC=C(C)C=C1)C1=CC=C(C)C=C1

Specifications

Quantity 200 mg
Formula Weight 1783.07
Physical Form Crystalline Powder
Chemical Name or Material [NH2Me2][(RuCl((S)-tolbinap))2(mu-Cl)3]
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.