Learn More
Cole-Parmer Cole-Parmer Replacement ionic strength adjustor, 2M (NH4)2SO4; 475 mL
Catalog No. 50633653 Shop All Cole-Parmer Products
Encompass_Preferred
Ensure consistent and accurate readings from your ISE with these ionic strength adjustors.
Catalog No. 50-633-653 Supplier Cole-Parmer Supplier No. 2750360
Ensure consistent and accurate readings from your ISE with these ionic strength adjustors.