Learn More
Strem, An Ascensus Company CAS# [15060-55-6]. 5g. Ammonium tetrathiomolybdate(VI), 99% (99.99%-Mo) PURATREM. MFCD00136013
SDP
Catalog No. 501979331
Encompass
CAS# 15060-55-6. 5g. Ammonium tetrathiomolybdate(VI), 99% (99.99%-Mo) PURATREM. MFCD00136013. Molecular Weight: 260.27. Molecular Formula: (NH4)2MoS4. Color/form: red-green xtl. Strem# 42-1000. www.strem.com/catalog/v3/42-1000/
Catalog No. 50-197-9331 Supplier Strem, An Ascensus Company Supplier No. 4210005G
CAS# 15060-55-6. 5g. Ammonium tetrathiomolybdate(VI), 99% (99.99%-Mo) PURATREM. MFCD00136013. Molecular Weight: 260.27. Molecular Formula: (NH4)2MoS4. Color/form: red-green xtl. Strem# 42-1000. www.strem.com/catalog/v3/42-1000/