Learn More
Lonza Walkersville B-ALI Bronchial Air-Liquid Interface Medium BulletKit
Catalog No. NC1769925
Encompass
B-ALI Bronchial Air Liquid Interface Medium BulletKit provides scientists with a unique tool to investigate new areas of airway epithelial research in a more physiologically relevant way. Content:1 x B-ALI Basal Media (00193516), 250 mL1 x B-ALI Differentiation Media (00193517), 500 mL1 x B-ALI SingleQuots Supplement Pack (00193515) containing:1 x Natural Cap Vial with Transferrin, 0.75 mL1 x Amber Vial with B-ALI-Inducer, 1.00 mL1 x Bottle BPE, 3.00 mL1 x Amber Vial with Epinephrine, 0.75 mL1 x Red Cap Vial with GA-1000, 0.75 mL1 x Green Cap Vial with hEGF, 0.75 mL1 x Natural Cap Vial with Hydrocortisone, 0.75 mL1 x Lilac Cap Vial with Insulin, 0.75 mL1 x Amber Vial with Retinoic Acid, 0.75 mL1 x Amber Vial with Triiodothyronine, 0.75 mL
Catalog No. NC1769925 Supplier Lonza Walkersville Supplier No. 00193514
B-ALI Bronchial Air Liquid Interface Medium BulletKit provides scientists with a unique tool to investigate new areas of airway epithelial research in a more physiologically relevant way. Content:1 x B-ALI Basal Media (00193516), 250 mL1 x B-ALI Differentiation Media (00193517), 500 mL1 x B-ALI SingleQuots Supplement Pack (00193515) containing:1 x Natural Cap Vial with Transferrin, 0.75 mL1 x Amber Vial with B-ALI-Inducer, 1.00 mL1 x Bottle BPE, 3.00 mL1 x Amber Vial with Epinephrine, 0.75 mL1 x Red Cap Vial with GA-1000, 0.75 mL1 x Green Cap Vial with hEGF, 0.75 mL1 x Natural Cap Vial with Hydrocortisone, 0.75 mL1 x Lilac Cap Vial with Insulin, 0.75 mL1 x Amber Vial with Retinoic Acid, 0.75 mL1 x Amber Vial with Triiodothyronine, 0.75 mL