Learn More
Genetex Inc Inhibin alpha Monoclonal Antibody [MSVA-561R] HistoMAX™ (GTX04415) GeneTex - IHC-Optimized Antibody.
SDP
Catalog No. NC3666876
Encompass_Preferred
The Inhibin alpha antibody [MSVA-561R] HistoMAX™ (GTX04415) is a Rabbit Monoclonal antibody in the GeneTex HistoMAX portfolio. The antibody is extensively evaluated for IHC-P using formalin-fixed human normal tissues (more than 70 different sections) and cancer tissue arrays to guarantee specificity and best-in-class performance. Information regarding both manual and autostainer staining protocols are available.Figure Legend: IHC-P analysis of human placenta tissue using GTX04415 Inhibin alpha antibody [MSVA-561R] HistoMAX. A strong inhibin alpha immunostaining is seen in the cyto and syncytiotrophoblast of the placenta of the first trimenon.
Catalog No. NC3666876 Supplier Genetex Inc Supplier No. GTX04415
The Inhibin alpha antibody [MSVA-561R] HistoMAX™ (GTX04415) is a Rabbit Monoclonal antibody in the GeneTex HistoMAX portfolio. The antibody is extensively evaluated for IHC-P using formalin-fixed human normal tissues (more than 70 different sections) and cancer tissue arrays to guarantee specificity and best-in-class performance. Information regarding both manual and autostainer staining protocols are available.Figure Legend: IHC-P analysis of human placenta tissue using GTX04415 Inhibin alpha antibody [MSVA-561R] HistoMAX. A strong inhibin alpha immunostaining is seen in the cyto and syncytiotrophoblast of the placenta of the first trimenon.