Learn More
Lonza Walkersville NHDC - Hu Dendritic Cells
Catalog No. 5000496339
Encompass
Dendritic Cells (DCs) act as the messenger cells of the immune system by processing antigens from pathogens and presenting the antigens to T cells to initiate an immune response Poietics Normal Human Dendritic Cells are tested for a panel of specific surface markers including CD11c CD86 CD80 HLADR and CD14- to provide detailed characterization of the cells These cells can survive up to 7 days in culture with the proper cytokines (IL-4 and GM-CSF) to maintain the cells as immature dendritic cells (iDCs) Cells can be further differentiated to mature dendritic cells with the addition of TNF- and IL-1 Cryopreserved Poietics Normal Human Dendritic Cells (NHDC) are guaranteed to contain 3 million viable cells per ampoule
Catalog No. 50-004-96339 Supplier Lonza Walkersville Supplier No. CC2701
Dendritic Cells (DCs) act as the messenger cells of the immune system by processing antigens from pathogens and presenting the antigens to T cells to initiate an immune response Poietics Normal Human Dendritic Cells are tested for a panel of specific surface markers including CD11c CD86 CD80 HLADR and CD14- to provide detailed characterization of the cells These cells can survive up to 7 days in culture with the proper cytokines (IL-4 and GM-CSF) to maintain the cells as immature dendritic cells (iDCs) Cells can be further differentiated to mature dendritic cells with the addition of TNF- and IL-1 Cryopreserved Poietics Normal Human Dendritic Cells (NHDC) are guaranteed to contain 3 million viable cells per ampoule