Learn More
Lonza Walkersville NHDF-Ad - Human Dermal Fibroblasts, Adult
Catalog No. NC9597467
Encompass
Human Dermal Fibroblasts are available from either adult skin (NHDF-Ad) or neonatal foreskin (NHDF-Neo). They are derived from the dermis of normal human neonatal foreskin or adult skin. They are cryopreserved at the end of primary culture. Normal Human Epidermal Keratinocytes (00192627) from matched donors are available for certain lots.Lonza's human dermal fibroblasts are guaranteed through 15 population doublings when using FGM-2 Growth Media. Dermal fibroblasts are characterized by morphological observation throughout serial passage.All cells test negative for mycoplasma, bacteria, yeast, and fungi. HIV-1, hepatitis B and hepatitis C are not detected for all donors and/or cell lots. A Certificate of Analysis is provided for each cell lot purchased.Applications:ECM protein analysisWound healingCollagen metabolismCosmeticsSkin therapy/models Content:1 x Cryopreserved ampule of NHDF-Ad containing ≥ 500,000 cells
Catalog No. NC9597467 Supplier Lonza Walkersville Supplier No. CC2511
Human Dermal Fibroblasts are available from either adult skin (NHDF-Ad) or neonatal foreskin (NHDF-Neo). They are derived from the dermis of normal human neonatal foreskin or adult skin. They are cryopreserved at the end of primary culture. Normal Human Epidermal Keratinocytes (00192627) from matched donors are available for certain lots.Lonza's human dermal fibroblasts are guaranteed through 15 population doublings when using FGM-2 Growth Media. Dermal fibroblasts are characterized by morphological observation throughout serial passage.All cells test negative for mycoplasma, bacteria, yeast, and fungi. HIV-1, hepatitis B and hepatitis C are not detected for all donors and/or cell lots. A Certificate of Analysis is provided for each cell lot purchased.Applications:ECM protein analysisWound healingCollagen metabolismCosmeticsSkin therapy/models Content:1 x Cryopreserved ampule of NHDF-Ad containing ≥ 500,000 cells