Learn More
Aviva Systems Biology Corp NHEJ1 Antibody - C-terminal region (ARP61619_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP61619_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against NHEJ1. It was validated on Western Blot. . Alternate names: FLJ12610, XLFApplicationWB. Species reactivity: Dog, Human, Rat
Catalog No. 50-287-423