Learn More
Promocell GmbH NHEK.F GROWTH MEDIUM 3
Catalog No. NC2220518
Encompass
PromoCell NHEK.f Pellet from > 1 million Normal Human Epidermal Keratinocytes, derived from foreskin, single donor cultured in Growth Medium 3
This item is not returnable. View return policy
Catalog No. NC2220518 Supplier Promocell GmbH Supplier No. C14046
PromoCell NHEK.f Pellet from > 1 million Normal Human Epidermal Keratinocytes, derived from foreskin, single donor cultured in Growth Medium 3