Learn More
Lonza Walkersville NHEK-Neo – Human Epidermal Keratinocytes, Neonatal, Pooled
Catalog No. NC2235577
Encompass
Cryopreserved Normal Human Epidermal Keratinocytes from pooled donros are guaranteed through 18 population doublings for adult and neonatal cells.All cells test negative for mycoplasma, bacteria, yeast, and fungi. HIV-1, hepatitis B and hepatitis C are not detected for all donors and/or cell lots. A Certificate of Analysis is provided for each cell lot purchased. Content:1 x Cryopreserved ampule of Human Neonatal Epidermal Keratinocytes containing ≥ 500,000 cells
This item is not returnable. View return policy
Catalog No. NC2235577 Supplier Lonza Walkersville Supplier No. 00192906
Cryopreserved Normal Human Epidermal Keratinocytes from pooled donros are guaranteed through 18 population doublings for adult and neonatal cells.All cells test negative for mycoplasma, bacteria, yeast, and fungi. HIV-1, hepatitis B and hepatitis C are not detected for all donors and/or cell lots. A Certificate of Analysis is provided for each cell lot purchased. Content:1 x Cryopreserved ampule of Human Neonatal Epidermal Keratinocytes containing ≥ 500,000 cells