Learn More
Promocell GmbH NHEM M2 AD PELLET 1MIO.CELLS
Supplier:  Promocell GmbH C14043
Encompass
Normal Human Epidermal Melanocytes, derived from adult skin, Cell-Pellet from 1 million cells dissolved in 200 μl RNAlater®.
Catalog No. 50-306-135