Learn More
Lonza Walkersville NHEM-Neo - Hu Epidermal Melano
Catalog No. 5000496403
Encompass
Adult and Neonatal Human Epidermal Melanocytes (NHEM) are the pigment producing cells located in the epidermal layers of all mammalian skin The function of the melanocyte is to produce melanin the pigment that gives skin its color Melanin protects skin from UV damage Skin is comprised of 2 main layers the epidermis and the dermis Our Cryopreserved Normal Human Epidermal Melanocytes are guaranteed through 8 population doublings express Mel-5 and L-Dopa conversion to melanin All cells test negative for mycoplasma bacteria yeast and fungi HIV-1 hepatitis B and hepatitis C are not detected for all donors and/or cell lots A Certificate of Analysis is provided for each cell lot purchased Note Melanocytes are extremely sensitive to over trypsinization Use Lonza s Trypsin/EDTA Solution (CC-5012) and dilute 1 1 with Versene (17-711E) prior to use
Catalog No. 50-004-96403 Supplier Lonza Walkersville Supplier No. CC2504
Add to Cart
Add to Cart
Adult and Neonatal Human Epidermal Melanocytes (NHEM) are the pigment producing cells located in the epidermal layers of all mammalian skin The function of the melanocyte is to produce melanin the pigment that gives skin its color Melanin protects skin from UV damage Skin is comprised of 2 main layers the epidermis and the dermis Our Cryopreserved Normal Human Epidermal Melanocytes are guaranteed through 8 population doublings express Mel-5 and L-Dopa conversion to melanin All cells test negative for mycoplasma bacteria yeast and fungi HIV-1 hepatitis B and hepatitis C are not detected for all donors and/or cell lots A Certificate of Analysis is provided for each cell lot purchased Note Melanocytes are extremely sensitive to over trypsinization Use Lonza s Trypsin/EDTA Solution (CC-5012) and dilute 1 1 with Versene (17-711E) prior to use