Learn More
Cayman Chemical NheptanoylLHomoserine lactone
Supplier: Cayman Chemical 100111985 MG
A small diffusible signaling molecule involved in quorum sensing, controlling gene expression, and affecting cellular metabolism; applications of this molecule include regulation of virulence, infection prevention, and septicemia in fish