Learn More
Cayman Chemical N-heptanoyl-L-Homoserine lactone, Purity: ≥98%, CAS: 177158-20-2, 10 mg
SDP
Catalog No. 502889470
Encompass_Preferred
A small diffusible signaling molecule involved in quorum sensing, controlling gene expression, and affecting cellular metabolism; applications of this molecule include regulation of virulence, infection prevention, and septicemia in fish
Catalog No. 50-288-9470 Supplier Cayman Chemical Supplier No. 1001119810 MG
A small diffusible signaling molecule involved in quorum sensing, controlling gene expression, and affecting cellular metabolism; applications of this molecule include regulation of virulence, infection prevention, and septicemia in fish