Learn More
Cayman Chemical NheptanoylLHomoserine lactone
SDP
Catalog No. 502887727
Encompass_Preferred
A small diffusible signaling molecule involved in quorum sensing, controlling gene expression, and affecting cellular metabolism; applications of this molecule include regulation of virulence, infection prevention, and septicemia in fish
Catalog No. 50-288-7727 Supplier Cayman Chemical Supplier No. 1001119850 MG
A small diffusible signaling molecule involved in quorum sensing, controlling gene expression, and affecting cellular metabolism; applications of this molecule include regulation of virulence, infection prevention, and septicemia in fish