Learn More
Cayman Chemical NhexadecnoylLHomoserIn l 10mg
SDP
Catalog No. 502954704
Encompass_Preferred
One of a number of lipophilic, long acyl side-chain bearing AHLs produced by the LuxI AHL synthase homolog SinI involved in quorum-sensing signaling in Sinorhizobium meliloti, a nitrogen-fixing bacterial symbiont of certain legumes; the most abundant AHL produced by the proteobacterium Rhodobacter capsulatus; activates genetic exchange between R. capsulatus cells
Catalog No. 50-295-4704 Supplier Cayman Chemical Supplier No. 1306410 MG
One of a number of lipophilic, long acyl side-chain bearing AHLs produced by the LuxI AHL synthase homolog SinI involved in quorum-sensing signaling in Sinorhizobium meliloti, a nitrogen-fixing bacterial symbiont of certain legumes; the most abundant AHL produced by the proteobacterium Rhodobacter capsulatus; activates genetic exchange between R. capsulatus cells