Learn More
Cayman Chemical N-hexadecanoyl-L-Homoserine lactone, Purity: ≥95%, CAS: 87206-01-7, 5 mg
SDP
Catalog No. 502953212
Encompass_Preferred
One of a number of lipophilic, long acyl side-chain bearing AHLs produced by the LuxI AHL synthase homolog SinI involved in quorum-sensing signaling in Sinorhizobium meliloti, a nitrogen-fixing bacterial symbiont of certain legumes; the most abundant AHL produced by the proteobacterium Rhodobacter capsulatus; activates genetic exchange between R. capsulatus cells
Catalog No. 50-295-3212 Supplier Cayman Chemical Supplier No. 130645 MG
One of a number of lipophilic, long acyl side-chain bearing AHLs produced by the LuxI AHL synthase homolog SinI involved in quorum-sensing signaling in Sinorhizobium meliloti, a nitrogen-fixing bacterial symbiont of certain legumes; the most abundant AHL produced by the proteobacterium Rhodobacter capsulatus; activates genetic exchange between R. capsulatus cells