Learn More
Cayman Chemical NhexanoylDLHomoserine lactone
SDP
Catalog No. 502890518
Encompass_Preferred
A bacterial quorum sensing signaling molecule; produced by mammalian pathogenic and rhizosphere-colonizing bacteria; application to tomato leaves suppresses the development of lesions induced by the plant pathogenic fungi B. cinerea at 60 µM; increases root length and shoot diameter when applied to A. thaliana seedlings in a hydroponic growth system at 10 µM
Catalog No. 50-289-0518 Supplier Cayman Chemical Supplier No. 100111975 MG
A bacterial quorum sensing signaling molecule; produced by mammalian pathogenic and rhizosphere-colonizing bacteria; application to tomato leaves suppresses the development of lesions induced by the plant pathogenic fungi B. cinerea at 60 µM; increases root length and shoot diameter when applied to A. thaliana seedlings in a hydroponic growth system at 10 µM