Learn More
Cayman Chemical N-hexanoyl-DL-Homoserine lactone, Purity: ≥98%, CAS: 106983-28-2, 10 mg
SDP
Catalog No. 502888350
Encompass_Preferred
A bacterial quorum sensing signaling molecule; produced by mammalian pathogenic and rhizosphere-colonizing bacteria; application to tomato leaves suppresses the development of lesions induced by the plant pathogenic fungi B. cinerea at 60 µM; increases root length and shoot diameter when applied to A. thaliana seedlings in a hydroponic growth system at 10 µM
Catalog No. 50-288-8350 Supplier Cayman Chemical Supplier No. 1001119710 MG
A bacterial quorum sensing signaling molecule; produced by mammalian pathogenic and rhizosphere-colonizing bacteria; application to tomato leaves suppresses the development of lesions induced by the plant pathogenic fungi B. cinerea at 60 µM; increases root length and shoot diameter when applied to A. thaliana seedlings in a hydroponic growth system at 10 µM