Learn More
Cayman Chemical NhexanoylLHomoserine lactone
SDP
Supplier:  Cayman Chemical 1000789625 MG
Encompass_Preferred
A bacterial quorum sensing signal molecule; regulates virulence in general and in cystic fibrosis, infection prevention, slime and biofilm reduction in commercial agriculture and aquaculture industries, food spoilage prevention, and septicemia in fish
Catalog No. 50-288-9089