Learn More
Cayman Chemical NhexanoylLHomoserine lactone
Supplier: Cayman Chemical 1000789650 MG
A bacterial quorum sensing signal molecule; regulates virulence in general and in cystic fibrosis, infection prevention, slime and biofilm reduction in commercial agriculture and aquaculture industries, food spoilage prevention, and septicemia in fish