Learn More
Cayman Chemical N-hexanoyl-L-Homoserine lactone, Purity: ≥95%, CAS: 147852-83-3, 10 mg
SDP
Catalog No. 502889527
Encompass_Preferred
A bacterial quorum sensing signal molecule; regulates virulence in general and in cystic fibrosis, infection prevention, slime and biofilm reduction in commercial agriculture and aquaculture industries, food spoilage prevention, and septicemia in fish
Catalog No. 50-288-9527 Supplier Cayman Chemical Supplier No. 1000789610 MG
A bacterial quorum sensing signal molecule; regulates virulence in general and in cystic fibrosis, infection prevention, slime and biofilm reduction in commercial agriculture and aquaculture industries, food spoilage prevention, and septicemia in fish