Learn More
Aviva Systems Biology Corp NHLRC2 Antibody - N-terminal region (ARP55889_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP55889_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against NHLRC2. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human NHLRC2. Alternate names: DKFZp779F115, FLJ20147, FLJ25621, FLJ33312, MGC45492ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rat, Zebrafish
Catalog No. 50-014-61649