Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NHP Compensation Set, Clone: L200, SP34-2, SK1, BD

Catalog No. BDB558640 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
20 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB558640 20 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB558640 Supplier BD Biosciences Supplier No. 558640
Add to Cart
Add to Cart

Flow Cytometry

TRUSTED_SUSTAINABILITY

Specifications

Conjugate FITC/APC/PE
Purification Method Affinity Purified
Quantity 20 Tests
Isotype IgG1 κ, IgG1 λ
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.