Learn More
Aviva Systems Biology Corp SNU13 Antibody - N-terminal region (ARP40236_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP40236_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against NHP2L1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human NHP2L1. Alternate names: NHPX, OTK27, SNU13, FA1, FA-1, 15.5K, SSFA1, SPAG12, SNRNP15-5ApplicationWB. Species reactivity: Cow, Dog, Goat, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Yeast, Zebrafish
Catalog No. 50-014-62960