Learn More
Medchemexpress LLC Nhpi-PEG3-C2-Pfp ester | 2101206-87-3 | MFCD28556928 | 533.40 g·mol⁻¹ | C23H20F5NO8 | 25 MG
SDP
Catalog No. 5000203155
Encompass_Preferred
NHPI-PEG3-C2-Pfp ester is a noncleavable three-unit polyethylene glycol (PEG) linker used in the synthesis of antibody-drug conjugates (ADCs). It acts as a short, hydrophilic spacer between payload and antibody, and is supplied for research use.
  • Noncleavable three-unit PEG linker.
  • Designed for antibody-drug conjugate synthesis.
  • Provides a hydrophilic spacer between payload and antibody.
  • Supplied for research use; consult the certificate of analysis for storage and purity details.
  • Available in multiple small pack sizes for method development and conjugation studies.
Catalog No. 50-002-03155 Supplier Medchemexpress LLC Supplier No. HY13009225MG
NHPI-PEG3-C2-Pfp ester is a noncleavable three-unit polyethylene glycol (PEG) linker used in the synthesis of antibody-drug conjugates (ADCs). It acts as a short, hydrophilic spacer between payload and antibody, and is supplied for research use.
  • Noncleavable three-unit PEG linker.
  • Designed for antibody-drug conjugate synthesis.
  • Provides a hydrophilic spacer between payload and antibody.
  • Supplied for research use; consult the certificate of analysis for storage and purity details.
  • Available in multiple small pack sizes for method development and conjugation studies.