Learn More
Medchemexpress LLC Nhpi-peg4-c2-nhs ester | 1415328-95-8 | MFCD28155511 | 98.9% | 508.48 | C23H28N2O11 | 50 MG
SDP
Catalog No. 5000202141
Encompass_Preferred
NHPI-PEG4-C2-NHS ester is an NHS-activated PEGylated linker reagent used in the synthesis of antibody-drug conjugates (ADCs). The molecule provides an activated ester for efficient coupling to primary amines and a PEG4 spacer that improves solubility and linker flexibility, supplied as a viscous liquid with high purity for research applications.
  • Activated NHS ester for rapid conjugation to primary amines.
  • Peg4 spacer improves aqueous solubility and linker flexibility.
  • Suitable for construction of antibody-drug conjugates.
  • High purity, viscous liquid form appropriate for research use.
  • Store at -20°C in pure form; stability is reduced in solvent.
Catalog No. 50-002-02141 Supplier Medchemexpress LLC Supplier No. HY13009150MG
NHPI-PEG4-C2-NHS ester is an NHS-activated PEGylated linker reagent used in the synthesis of antibody-drug conjugates (ADCs). The molecule provides an activated ester for efficient coupling to primary amines and a PEG4 spacer that improves solubility and linker flexibility, supplied as a viscous liquid with high purity for research applications.
  • Activated NHS ester for rapid conjugation to primary amines.
  • Peg4 spacer improves aqueous solubility and linker flexibility.
  • Suitable for construction of antibody-drug conjugates.
  • High purity, viscous liquid form appropriate for research use.
  • Store at -20°C in pure form; stability is reduced in solvent.