Learn More
Medchemexpress LLC NHPI-PEG4-C2-NHS ester | 1415328-95-8 | MFCD28155511 | 98.9% | 508.48 g/mol | C23H28N2O11 | 100 MG
SDP
Catalog No. 5000202268
Encompass_Preferred
NHPI-PEG4-C2-NHS ester is an activated N-hydroxyphthalimide (NHPI) N-hydroxysuccinimide (NHS) ester bearing a PEG4-C2 spacer, supplied as a research-grade ADC linker reagent for use in the synthesis of antibody-drug conjugates. The compound has CAS 1415328-95-8, molecular weight 508.48 g/mol, chemical formula C23H28N2O11, and a reported purity of 98.9%.
  • Activated NHS ester for conjugation to amines.
  • PEG4-C2 spacer provides hydrophilicity and distance between payload and antibody.
  • High reported purity suitable for research applications.
  • Available in multiple package sizes, including 100 mg.
  • Supplied with product data sheet and safety data sheet for handling information.
Catalog No. 50-002-02268 Supplier Medchemexpress LLC Supplier No. HY130091100MG
NHPI-PEG4-C2-NHS ester is an activated N-hydroxyphthalimide (NHPI) N-hydroxysuccinimide (NHS) ester bearing a PEG4-C2 spacer, supplied as a research-grade ADC linker reagent for use in the synthesis of antibody-drug conjugates. The compound has CAS 1415328-95-8, molecular weight 508.48 g/mol, chemical formula C23H28N2O11, and a reported purity of 98.9%.
  • Activated NHS ester for conjugation to amines.
  • PEG4-C2 spacer provides hydrophilicity and distance between payload and antibody.
  • High reported purity suitable for research applications.
  • Available in multiple package sizes, including 100 mg.
  • Supplied with product data sheet and safety data sheet for handling information.