Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Nhpi-PEG4-C2-Pfp ester | 2101206-46-4 | >97.0% | 577.45 g/mol | C25H24F5NO9 | 1 G
SDP

Catalog No. 5000203954
Encompass_Preferred

NHPI-PEG4-C2-Pfp ester is a PEG4-based linker bearing an N-hydroxyphthalimide (NHPI) activated pentafluorophenyl (Pfp) ester used for bioconjugation and the synthesis of antibody-drug conjugates (ADCs).

  • Used as a linker for antibody-drug conjugate synthesis.
  • Contains a non-cleavable four-unit PEG spacer for improved solubility.
  • Activated pentafluorophenyl ester enables efficient coupling to primary amines.
  • Purity greater than 97.0% as confirmed by certificate of analysis.
  • Molecular weight 577.45 g/mol and CAS number 2101206-46-4.

Catalog No. 50-002-03954 Supplier Medchemexpress LLC Supplier No. HY1300901G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

NHPI-PEG4-C2-Pfp ester is a PEG4-based linker bearing an N-hydroxyphthalimide (NHPI) activated pentafluorophenyl (Pfp) ester used for bioconjugation and the synthesis of antibody-drug conjugates (ADCs).

  • Used as a linker for antibody-drug conjugate synthesis.
  • Contains a non-cleavable four-unit PEG spacer for improved solubility.
  • Activated pentafluorophenyl ester enables efficient coupling to primary amines.
  • Purity greater than 97.0% as confirmed by certificate of analysis.
  • Molecular weight 577.45 g/mol and CAS number 2101206-46-4.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.