Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC NHPI-PEG4-C2-Pfp ester | 2101206-46-4 | MFCD28899317 | ≥97.0% | 577.45 g/mol | C25H24F5NO9 | 250 MG
SDP

Catalog No. 5000203961
Encompass_Preferred

NHPI-PEG4-C2-Pfp ester is a pentafluorophenyl-activated ester containing an N-hydroxyphthalimide-PEG4-C2 spacer. It is supplied as a high-purity research reagent used as a reactive linker intermediate for antibody-drug conjugate (ADC) synthesis and bioconjugation applications.

  • Purity ≥97.0% (NMR)
  • CAS number 2101206-46-4
  • Molecular weight 577.45 g/mol
  • Chemical formula C25H24F5NO9
  • Used as a reactive linker for antibody-drug conjugates and bioconjugation
  • Recommended storage: -20°C for up to 3 years; in solvent -80°C for up to 6 months

Catalog No. 50-002-03961 Supplier Medchemexpress LLC Supplier No. HY130090250MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

NHPI-PEG4-C2-Pfp ester is a pentafluorophenyl-activated ester containing an N-hydroxyphthalimide-PEG4-C2 spacer. It is supplied as a high-purity research reagent used as a reactive linker intermediate for antibody-drug conjugate (ADC) synthesis and bioconjugation applications.

  • Purity ≥97.0% (NMR)
  • CAS number 2101206-46-4
  • Molecular weight 577.45 g/mol
  • Chemical formula C25H24F5NO9
  • Used as a reactive linker for antibody-drug conjugates and bioconjugation
  • Recommended storage: -20°C for up to 3 years; in solvent -80°C for up to 6 months

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.