Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

G-Biosciences NHS activated Magnetic Beads
SCI_ED

Catalog No. S001131
Change view
Click to view available options
Capacity (Metric):
1 mL
5 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric)
S001131 1 mL
S001129 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S001131 Supplier G-Biosciences Supplier No. 7862216
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

NHS-Activated Magnetic Beads enable covalent, amine-based conjugation of proteins to magnetic beads for use in custom affinity purification.

  • The activated magnetic beads contain N-hydroxy-succinimide (NHS) functional groups that react with primary amines forming stable amide linkages.

Specifications

Particle Size 1 to 3 μm
Form Resin Slurry
Product Line DYNAL, DynaMag
For Use With (Application) Amine-based conjugation of proteins to magnetic beads for use in custom affinity purification
Type Magnetic Affinity Bead
Capacity (Metric) 1 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.