Learn More
Bioworld NHS-ACTIVATED SEPAROPOREA
Catalog No. 50197726
Encompass
NHS-ACTIVATED SEPAROPORE (AGAROSE) 4B-CL: NHS-activated Separopore 4B-CL Pre-activated medium for immobilization of ligands containing a primary amino group.; N-hydroxyl succinimide esters to form a stable amide (peptide) bond with the p...
Catalog No. 50-197-726 Supplier Bioworld Supplier No. 201810254
NHS-ACTIVATED SEPAROPORE (AGAROSE) 4B-CL: NHS-activated Separopore 4B-CL Pre-activated medium for immobilization of ligands containing a primary amino group.; N-hydroxyl succinimide esters to form a stable amide (peptide) bond with the p...