Learn More
Apexbio Technology LLC NHS-LC-LC-BIOTIN-KIT-2T
SDP
Catalog No. 5000567702
Encompass_Preferred
NHS-LC-LC-Biotin Kit is used in biochemical and molecular biology research originating from the need to label proteins and other macromolecules with high specificity It operates through NHS ester chemistry allowing biotin to covalently bind to primary amine groups in biological molecules facilitating subsequent detection or purification via biotin-avidin/streptavidin interactions The kit demonstrates robust in vitro activity in nanomolar to low micromolar ranges providing a reliable means for the modification of proteins under controlled conditions Typical applications include cell labeling immunoassays ELISA Western blotting and targeted drug delivery studies It is also employed in cell models and animal studies to track cellular processes or biomolecular interactions Due to its versatility the NHS-LC-LC-Biotin Kit is a staple in drug screening and various experimental setups with concentrations tailored to specific research designs
Catalog No. 50-005-67702 Supplier Apexbio Technology LLC Supplier No. K44012
NHS-LC-LC-Biotin Kit is used in biochemical and molecular biology research originating from the need to label proteins and other macromolecules with high specificity It operates through NHS ester chemistry allowing biotin to covalently bind to primary amine groups in biological molecules facilitating subsequent detection or purification via biotin-avidin/streptavidin interactions The kit demonstrates robust in vitro activity in nanomolar to low micromolar ranges providing a reliable means for the modification of proteins under controlled conditions Typical applications include cell labeling immunoassays ELISA Western blotting and targeted drug delivery studies It is also employed in cell models and animal studies to track cellular processes or biomolecular interactions Due to its versatility the NHS-LC-LC-Biotin Kit is a staple in drug screening and various experimental setups with concentrations tailored to specific research designs