Learn More
Bioworld NHS LC SPDP , 100 mg
Catalog No. 50199259
Encompass_Preferred
NHS LC SPDP, Succinimidyl 6-[3-(2-pyridylthio) propionamido] HexanoateSpacer arm SPDPAntibody-toxin conjugationReferences: 1.) Carlsson, J., et al. (1978) Biochem. J. 173, 723-737.M.W. = 425.5
Catalog No. 50-199-259 Supplier Bioworld Supplier No. 218100111
NHS LC SPDP, Succinimidyl 6-[3-(2-pyridylthio) propionamido] HexanoateSpacer arm SPDPAntibody-toxin conjugationReferences: 1.) Carlsson, J., et al. (1978) Biochem. J. 173, 723-737.M.W. = 425.5