Learn More
Sigma Aldrich Fine Chemicals Biosciences Nhs Mag SepharoseR Cytiva
Catalog No. 502900714
Encompass
NHS Mag Sepharose(R) are magnetic beads designed for pull-down techniques enabling rapid capture and enrichment of selected proteins based on affinity.
Catalog No. 50-290-0714 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. GE28951380
NHS Mag Sepharose(R) are magnetic beads designed for pull-down techniques enabling rapid capture and enrichment of selected proteins based on affinity.