Learn More
Medchemexpress LLC NHS-MMAF | 1404073-19-3 | 97.2% | 943.18 | 50 MG
SDP
Catalog No. 5000350103
Encompass_Preferred
NHS-MMAF is a modified MMAF extracted from patent WO2012143499, specifically intermediate 219. MMAF is a potent tubulin polymerization inhibitor and is used as an antitumor agent.
  • Modified MMAF.
  • Functions as a potent tubulin polymerization inhibitor.
  • Used as an antitumor agent.
  • Intended for research use only.
Catalog No. 50-003-50103 Supplier Medchemexpress LLC Supplier No. HY13932650MG
NHS-MMAF is a modified MMAF extracted from patent WO2012143499, specifically intermediate 219. MMAF is a potent tubulin polymerization inhibitor and is used as an antitumor agent.
  • Modified MMAF.
  • Functions as a potent tubulin polymerization inhibitor.
  • Used as an antitumor agent.
  • Intended for research use only.