Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC NHS-MMAF | 1404073-19-3 | 97.2% | 943.18 | C49H78N6O12 | 5 MG
SDP

Catalog No. 5000223037
Encompass_Preferred

NHS-MMAF is an NHS-activated derivative of monomethyl auristatin F (MMAF) designed as a linkerable payload for antibody-drug conjugate research. It acts as a tubulin polymerization inhibitor and is supplied as a solid for laboratory research use.

  • High purity suitable for research applications.
  • Contains NHS ester for conjugation to amines.
  • Potent tubulin polymerization inhibitor.
  • Soluble in DMSO; requires ultrasonic treatment and freshly opened DMSO.
  • Supplied as a solid; store at -20°C, protect from moisture and light.

Catalog No. 50-002-23037 Supplier Medchemexpress LLC Supplier No. HY1393265MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

NHS-MMAF is an NHS-activated derivative of monomethyl auristatin F (MMAF) designed as a linkerable payload for antibody-drug conjugate research. It acts as a tubulin polymerization inhibitor and is supplied as a solid for laboratory research use.

  • High purity suitable for research applications.
  • Contains NHS ester for conjugation to amines.
  • Potent tubulin polymerization inhibitor.
  • Soluble in DMSO; requires ultrasonic treatment and freshly opened DMSO.
  • Supplied as a solid; store at -20°C, protect from moisture and light.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.