Learn More
Medchemexpress LLC NHS-modified MMAF (trifluoroacetate) | 96.0% | 1165.34 | C57H87F3N8O14 | 50 MG
SDP
Catalog No. 5000203858
Encompass_Preferred
NHS-modified MMAF (trifluoroacetate) is an NHS-activated derivative of monomethyl auristatin F provided as the trifluoroacetate salt. It is used as a synthetic intermediate for preparing antibody-drug conjugate payload-linker constructs via NHS-ester-mediated conjugation to amines on antibody binders.
  • NHS-activated derivative for conjugation chemistry.
  • Provided as the trifluoroacetate salt for stability.
  • High purity suitable for research and ADC synthesis.
  • Available in research-scale quantities for lab use.
  • Documented with COA and SDS for quality and safety information.
Catalog No. 50-002-03858 Supplier Medchemexpress LLC Supplier No. HY139325A50MG
NHS-modified MMAF (trifluoroacetate) is an NHS-activated derivative of monomethyl auristatin F provided as the trifluoroacetate salt. It is used as a synthetic intermediate for preparing antibody-drug conjugate payload-linker constructs via NHS-ester-mediated conjugation to amines on antibody binders.
  • NHS-activated derivative for conjugation chemistry.
  • Provided as the trifluoroacetate salt for stability.
  • High purity suitable for research and ADC synthesis.
  • Available in research-scale quantities for lab use.
  • Documented with COA and SDS for quality and safety information.