Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC NHS-modified monomethyl auristatin F (TFA salt) | 1404073-10-4 | 96.0% | 1051.32 g/mol | C55H86N8O12 | 10 MG
SDP

Catalog No. 5000203946
Encompass_Preferred

NHS-Modified MMAF TFA is an NHS-activated derivative of monomethyl auristatin F provided as the trifluoroacetic acid (TFA) salt and used as a drug-linker intermediate for preparing antibody-drug conjugates (ADCs).

  • NHS-activated ester for efficient conjugation to primary amines
  • Provided as the TFA salt for isolation and handling
  • Intended as a drug-linker intermediate for ADC synthesis
  • Molecular formula C55H86N8O12; molecular weight 1051.32 g/mol
  • Commonly supplied in a 10 mg pack size

Catalog No. 50-002-03946 Supplier Medchemexpress LLC Supplier No. HY139325A10MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

NHS-Modified MMAF TFA is an NHS-activated derivative of monomethyl auristatin F provided as the trifluoroacetic acid (TFA) salt and used as a drug-linker intermediate for preparing antibody-drug conjugates (ADCs).

  • NHS-activated ester for efficient conjugation to primary amines
  • Provided as the TFA salt for isolation and handling
  • Intended as a drug-linker intermediate for ADC synthesis
  • Molecular formula C55H86N8O12; molecular weight 1051.32 g/mol
  • Commonly supplied in a 10 mg pack size

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.