Learn More
AdipoGen NHS Pacific Blue SE
SDP
Supplier:  AdipoGen CDXC0036M100
Encompass_Preferred
Chemical. CAS 215868-33-0. Formula C14H7F2NO7. MW 339.2. Synthetic. This amine-reactive reagent can be used to create blue-fluorescent bioconjugates with excitation/emission maxima ~410/455 nm that are excitable by the 405 nm spectral line of the blue diode violet laser. Used in highly sensitive analyses of amines, amino acids and dipeptides. Amine-reactive reagents react with non-protonated aliphatic amine groups, including the amine terminus of proteins and the є-amino group of lysines.
Catalog No. 50-256-1533