Learn More
Medchemexpress LLC HY-136304 10mg Medchemexpress, NHS-PEG1-SS-PEG1-NHS CAS:1688598-83-5 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY13630410MG
Encompass_Preferred
Medchemexpress, HY-136304 10mg NHS-PEG1-SS-PEG1-NHS CAS:1688598-83-5 NHS-PEG1-SS-PEG1-NHS is a reversible linker for biomacromolecule link with active small molecule. NHS-PEG1-SS-PEG1-NHS can be used in proteins liposomes or nanoparticles. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-203-0418