Learn More
Medchemexpress LLC NHS-PEG1-SS-PEG1-NHS | 1688598-83-5 | 98.0% | 436.41 | 50 MG
SDP
Catalog No. 5000348373
Encompass_Preferred
NHS-PEG1-SS-PEG1-NHS is a reversible linker designed for conjugating biomacromolecules with active small molecules.
  • Reversible linker
  • Designed for conjugating biomacromolecules with active small molecules
  • Suitable for use in proteins
  • Suitable for use in liposomes
  • Suitable for use in nanoparticles
Catalog No. 50-003-48373 Supplier Medchemexpress LLC Supplier No. HY13630450MG
NHS-PEG1-SS-PEG1-NHS is a reversible linker designed for conjugating biomacromolecules with active small molecules.
  • Reversible linker
  • Designed for conjugating biomacromolecules with active small molecules
  • Suitable for use in proteins
  • Suitable for use in liposomes
  • Suitable for use in nanoparticles