Learn More
Medchemexpress LLC Nhs-peg1-ss-peg1-nhs | 1688598-83-5 | MFCD32644554 | 98.0% | 436.41 | 100 MG
SDP
Catalog No. 5000348300
Encompass_Preferred
NHS-PEG1-SS-PEG1-NHS is a reversible linker designed for conjugating biomacromolecules with active small molecules. It is suitable for applications involving proteins, liposomes, or nanoparticles, and is intended for research use only.
  • Reversible linker for biomacromolecule conjugation
  • Can be used with active small molecules
  • Applicable in proteins, liposomes, or nanoparticles
  • For research use only
Catalog No. 50-003-48300 Supplier Medchemexpress LLC Supplier No. HY136304100MG
NHS-PEG1-SS-PEG1-NHS is a reversible linker designed for conjugating biomacromolecules with active small molecules. It is suitable for applications involving proteins, liposomes, or nanoparticles, and is intended for research use only.
  • Reversible linker for biomacromolecule conjugation
  • Can be used with active small molecules
  • Applicable in proteins, liposomes, or nanoparticles
  • For research use only