Learn More
Medchemexpress LLC NHS-PEG2-SS-PEG2-NHS | 96.7% | 580.63 g/mol | C22H32N2O12S2 | 5 MG
SDP
Catalog No. 5000202704
Encompass_Preferred
NHS-PEG2-SS-PEG2-NHS is a cleavable four-unit polyethylene glycol (PEG) linker with N-hydroxysuccinimide (NHS) ester end groups and a disulfide (SS) cleavable junction, supplied for use in antibody-drug conjugate synthesis and other bioconjugation applications.
  • Cleavable disulfide bond enables reductive release of payloads.
  • Hydrophilic PEG spacer improves solubility and reduces aggregation.
  • NHS ester end groups provide rapid amine coupling to proteins.
  • Moderate molecular weight supports spacer function without excessive steric bulk.
  • Available in small research sizes suitable for conjugation optimization.
Catalog No. 50-002-02704 Supplier Medchemexpress LLC Supplier No. HY1361335MG
NHS-PEG2-SS-PEG2-NHS is a cleavable four-unit polyethylene glycol (PEG) linker with N-hydroxysuccinimide (NHS) ester end groups and a disulfide (SS) cleavable junction, supplied for use in antibody-drug conjugate synthesis and other bioconjugation applications.
  • Cleavable disulfide bond enables reductive release of payloads.
  • Hydrophilic PEG spacer improves solubility and reduces aggregation.
  • NHS ester end groups provide rapid amine coupling to proteins.
  • Moderate molecular weight supports spacer function without excessive steric bulk.
  • Available in small research sizes suitable for conjugation optimization.