Learn More
Apexbio Technology LLC NHS-SS-11-Biotin(Synonyms: Sulfo-NHS-SS-Biotin, EZ-Link NHS-SS-Biotin, NHS-SS-D-Biotin, N-Hydroxysuccinimidyl-SS-Biotin), 5mg, CAS: 142439-92-7.
SDP
Catalog No. 5000060721
Encompass_Preferred
NHS-SS-11-Biotin (CAS 142439-92-7) is an amine-reactive biotinylation reagent commonly used in biochemical research Structurally it possesses an N-hydroxysuccinimide (NHS) ester group that covalently reacts with primary amines of target proteins or biomolecules forming stable amide bonds and releasing sulfo-NHS This reagent features a cleavable disulfide linker and an extended spacer arm of approximately 24 3 enhancing probe accessibility to avidin or streptavidin conjugates It is utilized extensively for reversible biotinylation affinity purification or labeling studies in proteomics and cell biology applications
Catalog No. 50-000-60721 Supplier Apexbio Technology LLC Supplier No. A80165
NHS-SS-11-Biotin (CAS 142439-92-7) is an amine-reactive biotinylation reagent commonly used in biochemical research Structurally it possesses an N-hydroxysuccinimide (NHS) ester group that covalently reacts with primary amines of target proteins or biomolecules forming stable amide bonds and releasing sulfo-NHS This reagent features a cleavable disulfide linker and an extended spacer arm of approximately 24 3 enhancing probe accessibility to avidin or streptavidin conjugates It is utilized extensively for reversible biotinylation affinity purification or labeling studies in proteomics and cell biology applications