Learn More
Apexbio Technology LLC NHS-SS-Biotin, 100mg. Cas: MFCD: MFCD00466919. Amine-reactive biotinylation reagent, mid-length
SDP
Catalog No. 501011217
Encompass_Preferred
Biotinyltation reagent used to attach biotin to primary amines, cleavage dislfide bond included for reversible labeling 142439-92-7. MFCD00466919
Catalog No. 50-101-1217 Supplier Apexbio Technology LLC Supplier No. A8006100
Biotinyltation reagent used to attach biotin to primary amines, cleavage dislfide bond included for reversible labeling 142439-92-7. MFCD00466919