Learn More
Apexbio Technology LLC NHS-SS-Biotin, 1g. Cas: MFCD: MFCD00466919. Amine-reactive biotinylation reagent, mid-length
SDP
Supplier:  Apexbio Technology LLC A80061000
Encompass_Preferred
NHS-SS-biotin (Succinimidyl-2-(biotinamido)-ethyl-1,3-dithiopropionate) is a long-chain cleavable amine-reactive biotinylation reagent that has a spacer arm of 24.3 A in length due to the existence of a cross-bridge within its chemical structure. Other sizes are available. Please inqury us for quote.
Catalog No. 50-190-4976