Learn More
Bioworld NHS Bromoacetate, 1 g
Supplier:  Bioworld 218100091
Encompass_Preferred
NHS Bromoacetate, N-Hydroxysuccinimidyl BromoacetatePrimary amine and sulfhydrylProtein-protein conjugationBernatowicz, M.S. and Matsueda, G.R. (1986) Anal. Biochem. 155, 95. 2.) Cuatrecasas, P., et al. (1969) J. Biol. Chem. 244, 4316.
Catalog No. 50-199-257